missing translation for 'onlineSavingsMsg'
Learn More

Felbinac Ethyl Ester, Mikromol™

Product Code. 30051058
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051058 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051058 Supplier Mikromol Supplier No. MM3501.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Felbinac Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Felbinac Ethyl Ester
CAS 14062-23-8
Analyte or Component Names Felbinac Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H16 O2
Certifications/Compliance ISO 9001
Synonym [1,1'-Biphenyl]-4-acetic acid ethyl ester, 4-Biphenylacetic acid, ethyl ester (8CI), Acetic acid, (4-biphenylyl)-, ethyl ester (7CI), Diatec, Ethyl 4-biphenylylacetate, Ethyl p-biphenylylacetate, Felbinac ethyl ester, LM 001
InChI Formula InChI=1S/C16H16O2/c1-2-18-16(17)12-13-8-10-15(11-9-13)14-6-4-3-5-7-14/h3-11H,2,12H2,1H3
SMILES CCOC(=O)Cc1ccc(cc1)c2ccccc2
IUPAC Name ethyl 2-(4-phenylphenyl)acetate
Molecular Weight (g/mol) 240.3
Quantity 100 mg
Formula Weight 240.115 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.