missing translation for 'onlineSavingsMsg'
Learn More

Felbamate, TRC

Product Code. 30008661
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008661 25 mg 25mg
30006751 250 mg 250mg
18090410 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008661 Supplier Toronto Research Chemicals Supplier No. TRCF23100025MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25451-15-4
Molecular Formula C11 H14 N2 O4
Molecular Weight (g/mol) 238.24
Synonym Felbamate
IUPAC Name (3-carbamoyloxy-2-phenylpropyl) carbamate
SMILES NC(=O)OCC(COC(=O)N)c1ccccc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H14N2O4/c12-10(14)16-6-9(7-17-11(13)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,12,14)(H2,13,15)
Formula Weight 238.0954
Percent Purity >95%
Chemical Name or Material Felbamate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.