missing translation for 'onlineSavingsMsg'
Learn More

Felbamate, MedChemExpress

Product Code. 16444277
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444277 50 mg 50mg
16488156 100 mg 100mg
16438388 10 mg 10mg
16431396 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444277 Supplier Medchem Express Supplier No. HYB0184
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Felbamate (W-554) is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA).

Felbamate (W-554) is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA).

Specifications

Chemical Name or Material Felbamate
Physical Form Solid
CAS 25451-15-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H14N2O4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym W-554 ADD-03055
Solubility Information DMSO : ≥ 100 mg/mL (419.74 mM)
SMILES NC(OCC(C1=CC=CC=C1)COC(N)=O)=O
Molecular Weight (g/mol) 238.24
Formula Weight 238.24
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.