missing translation for 'onlineSavingsMsg'
Learn More

Felbamate, MedChemExpress

Product Code. 16438388
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16438388

Brand: Medchem Express HYB0184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Felbamate (W-554) is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA).

Felbamate (W-554) is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA).

Specifications

Chemical Name or Material Felbamate
Physical Form Solid
CAS 25451-15-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H14N2O4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym W-554 ADD-03055
Solubility Information DMSO : ≥ 100 mg/mL (419.74 mM)
SMILES NC(OCC(C1=CC=CC=C1)COC(N)=O)=O
Molecular Weight (g/mol) 238.24
Formula Weight 238.24
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.