missing translation for 'onlineSavingsMsg'
Learn More

Felbamate-d4, TRC

Product Code. 30394818
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394818 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394818 Supplier Toronto Research Chemicals Supplier No. TRCF23100250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Felbamate-d4
CAS 106817-52-1
Purity Grade Notes HPLC
Molecular Formula C11 H10 D4 N2 O4
Quantity 50 mg
Synonym Felbamate-d4, Felbamyl-d4, Felbatol-d4, Taloxa-d4, CTK8F9733, ADD-03055-d4, W-554-d4, 2-Phenyl-1,3-propanedioldicarbamate-d4, Carbamic Acid 2-Phenyltrimethylene-d4 Ester
InChI Formula InChI=1S/C11H14N2O4/c12-10(14)16-6-9(7-17-11(13)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,12,14)(H2,13,15)/i6D2,7D2
SMILES [2H]C([2H])(OC(=O)N)C(c1ccccc1)C([2H])([2H])OC(=O)N
Molecular Weight (g/mol) 242.267
Formula Weight 242.1205
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.