missing translation for 'onlineSavingsMsg'
Learn More

Fedratinib, MedChemExpress

Product Code. p-200099793
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
This item is not returnable. View return policy

Product Code. 16443958

Brand: Medchem Express HY10409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fedratinib (TG-101348) is a potent, selective, ATP-competitive and orally active JAK2 inhibitor with IC50s of 3 nM for both JAK2 and JAK2V617F kinase. Fedratinib shows 35- and 334-fold selectivity over JAK1 and JAK3, respectively. Fedratinib induces cancer cell apoptosis and has the potential for myeloproliferative disorders research.

Fedratinib (TG-101348) is a potent, selective, ATP-competitive and orally active JAK2 inhibitor with IC50s of 3 nM for both JAK2 and JAK2V617F kinase. Fedratinib shows 35- and 334-fold selectivity over JAK1 and JAK3, respectively. Fedratinib induces cancer cell apoptosis and has the potential for myeloproliferative disorders research.

Specifications

Chemical Name or Material Fedratinib
CAS 936091-26-8
Color Light Nude
Purity Grade Notes Research
Molecular Formula C27H36N6O3S
Quantity 10 mg
Synonym TG-101348 SAR 302503
Solubility Information DMSO : 125 mg/mL (238.24 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC(NC2=NC(NC3=CC=C(C=C3)OCCN4CCCC4)=NC=C2C)=C1)(NC(C)(C)C)=O
Molecular Weight (g/mol) 524.68
Formula Weight 524.68
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.