missing translation for 'onlineSavingsMsg'
Learn More

Fedovapagon, MedChemExpress

Product Code. 16496238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496238 10 mg 10mg
16472367 50 mg 50mg
16428837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496238 Supplier Medchem Express Supplier No. HY14887
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fedovapagon is a selective vasopressin V2 receptor (V2R) agonist with an EC50 of 24 nM, which is being developed for the treatment of nocturia.

Fedovapagon is a selective vasopressin V2 receptor (V2R) agonist with an EC50 of 24 nM, which is being developed for the treatment of nocturia.

Specifications

Chemical Name or Material Fedovapagon
CAS 347887-36-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H34N4O3
Quantity 10 mg
Synonym VA106483
Solubility Information DMSO : ≥ 125 mg/mL (270.22 mM)
SMILES CC1=C(CNC(N2CCC[C@]2(C(N(C)C)=O)[H])=O)C=CC(C(N3CCCCC4=C3C=CC=C4)=O)=C1
Molecular Weight (g/mol) 462.58
Formula Weight 462.58
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.