missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FECH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14466-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is localized to the mitochondrion, where it catalyzes the insertion of the ferrous form of iron into protoporphyrin IX in the heme synthesis pathway. Mutations in this gene are associated with erythropoietic protoporphyria. Two transcript variants encoding different isoforms have been found for this gene. A pseudogene of this gene is found on chromosome 3.Specifications
| FECH | |
| Polyclonal | |
| Unconjugated | |
| FECH | |
| EPP, FCE, FECH, Ferrochelatase, mitochondrial, Heme synthase, Protoheme ferro lyase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14151, 2235, 361338 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22315, P22830 | |
| Fech | |
| FECH Fusion Protein Ag5814 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction