missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FECH Polyclonal specifically detects FECH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | FECH |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P22830 |
| Gene Alias | EC 4.99.1.1, EPP, FCE, ferrochelatase, ferrochelatase (protoporphyria), ferrochelatase, mitochondrial, Heme synthase, heme synthetase, Protoheme ferro-lyase, protoporphyria |
| Gene Symbols | FECH |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?