missing translation for 'onlineSavingsMsg'
Learn More

Febuxostat 67M-1, TRC

Product Code. 30396399
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396399 25 mg
30406752 10 mg
30389928 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396399 Supplier Toronto Research Chemicals Supplier No. TRCF22900525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887945-96-2
Molecular Formula C16H16N2O4S
Molecular Weight (g/mol) 332.37
Synonym 2-[3-Cyano-4-(3-hydroxy-2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid
SMILES OCC(C)COC1=C(C#N)C=C(C2=NC(C)=C(C(O)=O)S2)C=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16N2O4S/c1-9(7-19)8-22-13-4-3-11(5-12(13)6-17)15-18-10(2)14(23-15)16(20)21/h3-5,9,19H,7-8H2,1-2H3,(H,20,21)
Formula Weight 332.0831
Chemical Name or Material Febuxostat 67M-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.