missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FDX1 Antibody (1E7), Novus Biologicals™

Product Code. 18347677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347677 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347677 Supplier Novus Biologicals™ Supplier No. H00002230M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FDX1 Monoclonal antibody specifically detects FDX1 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDX1
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 1E7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004100
Gene Alias Adrenal ferredoxin, adrenodoxin, mitochondrial, ADXhepatoredoxin, FDX, ferredoxin 1, ferredoxin-1, Hepatoredoxin, LOH11CR1D, mitochondrial adrenodoxin
Host Species Mouse
Immunogen FDX1 (NP_004100.1, 85 a.a. ∽ 183 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VGDSLLDVVVENNLDIDGFGACEGTLACSTCHLIFEDHIYEKLDAITDEENDMLDLAYGLTDRSRLGCQICLTKSMDNMTVRVPETVADARQSIDVGKT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2230
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.