missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDPS Rabbit anti-Mouse, Clone: 115, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90812-100UL
This item is not returnable.
View return policy
Description
FDPS Monoclonal specifically detects FDPS in Mouse samples. It is validated for ELISA.
Specifications
| FDPS | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 115 | |
| ELISA 1:5000-1:10000 | |
| 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293 | |
| Recombinant Mouse FDPS Protein | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 2224 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction