missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FDPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597742
This item is not returnable.
View return policy
Description
FDPS Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
The isoprene biosynthetic pathway supply the cell with cholesterol, ubiquinone, and various nonsterol metabolites. The farnesylpyrophosphate synthetase enzyme catalyzes the formation of geranyl and farnesylpyrophosphate from isopentenylpyrophosphate and dimethylallyl pyrophosphate. Analysis of FDPS activity and protein in rat liver, accompanied by immunofluorescence and immunoelectron microscopy studies, demonstrated that FDPS is predominantly localized in peroxisomes. 1 Liver tissue from patients with the peroxisomal deficiency diseases Zellweger syndrome and neonatal adrenoleukodystrophy exhibit diminished activities of FDPS and subsequent isoprenoid synthesis.
Specifications
| FDPS | |
| Polyclonal | |
| Unconjugated | |
| FDPS | |
| (2E,6E)-farnesyl diphosphate synthase; 6030492I17Rik; Ac2-125; AI256750; cholesterol-regulated 39 kDa protein; CR 39; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase); farnesyl diphosphate synthetase; farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; Fdps; Fdpsl1; FPP synthase; FPP synthetase; FPPS; FPS; geranyltranstransferase; KIAA1293; mKIAA1293; POROK9; testis-specific farnesyl pyrophosphate synthetase | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2224 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P14324 | |
| FDPS | |
| Recombinant Human Farnesyl pyrophosphate synthase protein (1-419AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction