missing translation for 'onlineSavingsMsg'
Learn More

FDPS Antibody (005), Biotin, Novus Biologicals™

Product Code. 18349930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349930 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349930 Supplier Novus Biologicals Supplier No. NBP290198B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FDPS Monoclonal specifically detects FDPS in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDPS
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 5
Conjugate Biotin
Formulation PBS
Gene Alias 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FDPS (NP_001129294.1; Met1-Lys353).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2224
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.