missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDFT1 Rabbit anti-Human, Mouse, Rat, Clone: 4W10J4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | FDFT1 |
|---|---|
| Clone | 4W10J4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326506
|
Novus Biologicals
NBP3-16456-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18332335
|
Novus Biologicals
NBP3-16456-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FDFT1 Monoclonal antibody specifically detects FDFT1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| FDFT1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2222 | |
| IgG | |
| Affinity purified |
| 4W10J4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DGPT, EC 2.5.1.21, ERG9, Farnesyl-diphosphate farnesyltransferase, farnesyl-diphosphate farnesyltransferase 1, FPP:FPP farnesyltransferase, presqualene-di-diphosphate synthase, SQS, squalene synthase, squalene synthetase, SS | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human FDFT1 (P37268). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title