missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDC Antibody (CNA.42), Novus Biologicals™
Mouse Monoclonal Antibody
£201.00 - £405.00
Specifications
| Antigen | FDC |
|---|---|
| Clone | CNA.42 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18265235
|
Novus Biologicals
NBP1-43118-0.025MG |
0.025 mg |
£201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790594
|
Novus Biologicals
NBP1-43118-0.1mg |
0.1 mg |
£405.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FDC Monoclonal specifically detects FDC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| FDC | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 1218 | |
| The immunogen for this antibody was follicular dendritic cells. | |
| Primary | |
| Store at 4C. Do not freeze. |
| CNA.42 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Human | |
| cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC | |
| CMD1B | |
| IgM | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title