missing translation for 'onlineSavingsMsg'
Learn More

fd/M13 bacteriophage Antibody (MM05), Alexa Fluor™ 350 , Novus Biologicals™

Product Code. 18426839 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426839 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426839 Supplier Novus Biologicals Supplier No. NBP305788AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

fd/M13 bacteriophage Monoclonal antibody specifically detects fd/M13 bacteriophage in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen fd/M13 bacteriophage
Applications ELISA
Classification Monoclonal
Clone MM05
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias major coat protein
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with M13 Bacteriophage.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 927333
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.