Learn More
FCRN/FCGRT Antibody (CL3638) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61626AF750
Product Code. 30292039
  • £558.00 / 0.10mL
Description
FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FCRN/FCGRT
Monoclonal
Alexa Fluor 750
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
This antibody was developed using a recombinant protein derived from P55899, with the exact immunogen sequence remaining proprietary.
0.1 mL
Immunology
2217
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3638
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified