missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17157-1-AP-150UL
This item is not returnable.
View return policy
Description
FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions.Specifications
| FCRLB | |
| Polyclonal | |
| Unconjugated | |
| FCRLB | |
| Fc receptor like B, Fc receptor like protein 2, Fc receptor related protein Y, FCRL2, FCRLB, FCRLM2, FCRLY, FcRY, FREB 2, FREB2, RP11 474I16.6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127943, 435653, 498275 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5DRQ8, Q6BAA4 | |
| FCRLB | |
| FCRLB Fusion Protein Ag10356 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction