missing translation for 'onlineSavingsMsg'
Learn More

FCRLA/FCRLM1 Antibody [PE], Novus Biologicals™

Product Code. 30734269 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734269 Supplier Novus Biologicals Supplier No. AF7905P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FCRLA/FCRLM1 Polyclonal antibody specifically detects FCRLA/FCRLM1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRLA/FCRLM1
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREBFc receptor-like and mucin-like protein 1, MGC4595
Host Species Sheep
Immunogen E. coli-derived recombinant mouse FCRLA, Lys221-Lys352 (Ile321Met), Accession # Q920A9
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84824
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.