missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FCRL5/FcRH5/IRTA2 Antibody (cevostamab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28621-1mg
This item is not returnable.
View return policy
Description
FCRL5/FcRH5/IRTA2 Monoclonal antibody specifically detects FCRL5/FcRH5/IRTA2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FCRL5/FcRH5/IRTA2 | |
| Monoclonal | |
| Unconjugated | |
| Q96RD9 | |
| Human | |
| Protein A purified | |
| RUO | |
| 83416 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| cevostamab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BXMAS1MGC119592, CD307, CD307e antigen, DKFZp667E2019, DKFZp667F216, Fc receptor homolog 5, Fc receptor-like 5, Fc receptor-like protein 5, FcRH5, FCRH5MGC119593, FcRL5, FcR-like protein 5, FLJ00333, FLJ00397, Immune receptor translocation-associated protein 2, immunoglobulin superfamily receptor translocation associated 2 (IRTA2), IRTA2MGC119590, PRO820 | |
| FcRH5/ IRTA2/ CD307e | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction