missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL5/FcRH5/IRTA2 Antibody (1010552), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB20872MFV610
This item is not returnable.
View return policy
Description
FCRL5/FcRH5/IRTA2 Monoclonal antibody specifically detects FCRL5/FcRH5/IRTA2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| FCRL5/FcRH5/IRTA2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 1010552 | |
| Flow Cytometry, CyTOF-ready | |
| BXMAS1MGC119592, CD307, CD307e antigen, DKFZp667E2019, DKFZp667F216, Fc receptor homolog 5, Fc receptor-like 5, Fc receptor-like protein 5, FcRH5, FCRH5MGC119593, FcRL5, FcR-like protein 5, FLJ00333, FLJ00397, Immune receptor translocation-associated protein 2, immunoglobulin superfamily receptor translocation associated 2 (IRTA2), IRTA2MGC119590, PRO820 | |
| Mouse myeloma cell line, NS0-derived human FCRL5 protein, Gln16-Arg844, Accession # Q96RD9 | |
| RUO | |
| 83416 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction