missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL4/FcRH4/IRTA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69383
This item is not returnable.
View return policy
Description
FCRL4/FcRH4/IRTA1 Polyclonal specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for Western Blot.
Specifications
| FCRL4/FcRH4/IRTA1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1MGC150523, MGC150522 | |
| Rabbit | |
| 57 kDa | |
| 100 μL | |
| Primary | |
| Porcine: 85%. | |
| Human, Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96PJ5 | |
| FCRL4 | |
| Synthetic peptides corresponding to FCRL4(Fc receptor-like 4) The peptide sequence was selected from the N terminal of FCRL4. Peptide sequence FKGERVTLTCNGFQFYATEKTTWYHRHYWGEKLTLTPGNTLEVRESGLYR. | |
| Affinity purified | |
| RUO | |
| 83417 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction