Learn More
FCRL4/FcRH4/IRTA1 Antibody (A1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11836AF594
Product Code. 18310552
  • £454.00 / 0.10mL
Description
FCRL4/FcRH4/IRTA1 Monoclonal antibody specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FCRL4/FcRH4/IRTA1
Monoclonal
Alexa Fluor 594
CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1MGC150523, MGC150522
DNA-immunization followed by a boost with FCRL4/FcRH4/IRTA1-transfected cells
0.1 mL
Primary
Human
Purified
Flow Cytometry
A1
50mM Sodium Borate
Mouse
Protein A purified
RUO
83417
Store at 4°C in the dark.
IgG2a κ