missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCRL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543525
This item is not returnable.
View return policy
Description
Peptide sequence: RGSSLSDAVH VEFSPDWLIL QALHPVFEGD NVILRCQGKD NKNTHQKVYY Sequence homology: Human: 100%.
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
Specifications
| FCRL3 | |
| Polyclonal | |
| Unconjugated | |
| FCRL3 | |
| BXMAS1-like protein 2; CD307c; CD307e; Fc receptor homolog 3; Fc receptor like 3; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRL3; Fcrl5; fcR-like protein 3; FcR-like protein 5; hIFGP3; IFGP family protein 3; IFGP3; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; mBXMH2; moFcRH3; rCG62831-like; SH2 domain-containing phosphatase anchor protein 2; SPAP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 115352 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96P31 | |
| FCRL3 | |
| Synthetic peptide directed towards the middle region of human FCRL3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction