missing translation for 'onlineSavingsMsg'
Learn More

FCRL3/FcRH3 Antibody, Novus Biologicals™

Product Code. 18431279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431279 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431279 Supplier Novus Biologicals Supplier No. H00115352B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

FCRL3/FcRH3 Polyclonal antibody specifically detects FCRL3/FcRH3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL3/FcRH3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. ABM86598.1
Gene Alias CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FCRH3, fcR-like protein 3, hIFGP3, IFGP3, immunoglobulin superfamily receptor translocation associated protein 3, IRTA3, SPAP2
Host Species Mouse
Immunogen FCRL3 (ABM86598.1, 1 a.a. - 742 a.a.) full-length human protein. MLLWLLLLILTPGREQSGVAPKAVLLLNPPWSTAFKGEKVALICSSISHSLAQGDTYWYHDEKLLKIKHDKIQITEPGNYQCKTRGSSLSDAVHVEFSPDWLILQALHPVFEGDNVILRCQGKDNKNTHQKVYYKDGKQLPNSYNLEKITVNSVSRDNSKYHCTAYRKFYILDIEVTSKPLNIQVQELFLHPVLRASSSTPIEGSPMTLTCETQLSPQRPDVQLQFSLFRDSQTLGLGWSRSPRLQIPAMWTEDSGSYWCEVETVTHSIKKRSLRSQIRVQRVPVSNVNLEIRPTGGQLIEGENMVLICSVAQGSGTVTFSWHKEGRVRSLGRKTQRSLLAELHVLTVKESDAGRYYCAADNVHSPILSTWIRVTVRIPVSHPVLTFRAPRAHTVVGDLLELHCESLRGSPPILYRFYHEDVTLGNSSAPSGGGASFNLSLTAEHSGNYSCDADNGLGAQHSHGVSLRVTVPVSRPVLTLRAPGAQAVVGDLLELHCESLRGSFPILYWFYHEDDTLGNISAHSGGGASFNLSLTTEHSGNYSCEADNGLGAQHSKVVTLNVTGTSRNRTGLTAAGITGLVLSILVLAAAAALLHYARARRKPGGLSATGTSSHSPSECQEPSSSRPSRIDPQEPTHSKPLAPMELEPMYSNVNPGDSNPIYSQIWSIQHTKENSANCPMMHQEHEELTVLYSELKKTHPDDSAGEASSRGRAHEEDDEENYENILNPRKNKVQDFPCLCNT
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers
Primary or Secondary Primary
Gene ID (Entrez) 115352
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.