missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCRL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109719
This item is not returnable.
View return policy
Description
Immunogen sequence: LTLVAPSSVF EGDSIVLKCQ GEQNWKIQKM AYHKDNKELS VFKKFSDFLI QSAVLSDSGN YFCSTKGQLF LWDKTSNIVK I.
The FCRL2 gene encodes the protein Fc receptor-like 2, which is part of the Fc receptor-like family involved in immune system regulation. FCRL2 is primarily expressed on the surface of B lymphocytes, where it contributes to the modulation of B cell activation and immune responses. As a member of the Fc receptor-like family, FCRL2 is thought to play roles in signaling pathways that influence B cell development, differentiation, and antibody production. It may act as a regulatory receptor that balances immune activation and inhibition, thereby contributing to immune homeostasis and prevention of autoimmunity. The expression of FCRL2 can impact conditions such as autoimmune diseases and B cell-related disorders, where dysregulation of B cell signaling is evident. Research into FCRL2 is ongoing to explore its specific ligand interactions and functional mechanisms, aiming to understand its potential as a therapeutic target for modulating immune responses in diseases characterized by B cell dysfunction.
Specifications
| FCRL2 | |
| Polyclonal | |
| Unconjugated | |
| FCRL2 | |
| CD307b; Fc receptor homolog 2; Fc receptor like 2; Fc receptor-like 2; Fc receptor-like protein 2; FCRH2; FCRL2; FcR-like protein 2; IFGP family protein 4; IFGP4; immune receptor translocation-associated protein 4; immunoglobulin receptor translocation-associated protein 4; immunoglobulin superfamily Fc receptor, gp42; IRTA4; RP11-367J7.6; SH2 domain containing phosphatase anchor protein 1; SH2 domain-containing phosphatase anchor protein 1; SPAP1; SPAP1A; SPAP1B; SPAP1C; UNQ9236/PRO31998 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79368 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96LA5 | |
| FCRL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-100 of human FCRL2 (Q96LA5). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction