missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCRL1 Monoclonal Antibody (8F7F2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548800
This item is not returnable.
View return policy
Description
FCRL1 Monoclonal Antibody for Western Blot, Flow, ELISA
Fc receptor-like 1 (FCRL1) is a gene situated on chromosome 1q23, encoding a protein that is part of the Fc receptor-like family, which shares structural similarities with classical Fc receptors involved in immune responses. FCRL1 is predominantly expressed in B lymphocytes and is thought to play a role in the regulation of immune functions, particularly those associated with B cell development and differentiation. Although its precise biological function is not fully understood, it is believed to act as an immunomodulatory receptor that could influence B cell signaling pathways. The expression patterns of FCRL1 suggest possible involvement in autoimmune disorders and immune system modulation, potentially contributing to pathological conditions when dysregulated. Research into FCRL1 and other Fc receptor-like proteins continues to explore their contributions to immune homeostasis and potential as therapeutic targets in diseases involving B cell dysfunction.
Specifications
| FCRL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q96LA6 | |
| Fcrl1 | |
| Purified recombinant fragment of human CD307A (AA: extra 17-202) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Western Blot | |
| 8F7F2 | |
| Unconjugated | |
| Fcrl1 | |
| A230020G22Rik; Bxmas1; BXMAS1-like protein 1; BXMAS1-like protein 1b; Bxmh1; Bxmh1b; CD307a; CD307a antigen; fc receptor 1-like protein; fc receptor homolog 1; Fc receptor like 1; Fc receptor-like 1; Fc receptor-like protein 1; Fcrh1; FCRH1DKFZp667O1421; Fcrh1l; Fcrh1s; Fcrl1; FcR-like protein 1; hIFGP1; IFGP family protein 1; IFGP1; immune receptor translocation-associated protein 5; immunoglobulin superfamily Fc receptor, gp42; IRTA5; mBXMH1; mIFGP1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 115350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction