missing translation for 'onlineSavingsMsg'
Learn More

FCRL1/FcRH1 Antibody [Biotin], Novus Biologicals™

Product Code. 30738079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30738079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738079 Supplier Novus Biologicals Supplier No. AF7286B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FCRL1/FcRH1 Polyclonal antibody specifically detects FCRL1/FcRH1 in Mouse samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL1/FcRH1
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5immunoglobulin superfamily Fc receptor, gp42
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FCRL1/FcRH1, Ala17-Ser219, Accession # Q8R4Y0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115350
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.