missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL1/FcRH1 Antibody (282415), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2049MFV500
This item is not returnable.
View return policy
Description
FCRL1/FcRH1 Monoclonal antibody specifically detects FCRL1/FcRH1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| FCRL1/FcRH1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 282415 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5immunoglobulin superfamily Fc receptor, gp42 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL1/FcRH1, Ala17-His304, Accession # Q96LA6 | |
| RUO | |
| 115350 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction