missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCN3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11867-1-AP-150UL
This item is not returnable.
View return policy
Description
H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.Specifications
| FCN3 | |
| Polyclonal | |
| Unconjugated | |
| FCN3 | |
| FCN3, FCNH, Ficolin 3, HAKA1, Hakata antigen | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8547 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75636 | |
| FCN3 | |
| FCN3 Fusion Protein Ag2449 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction