missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCHSD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12654-50ul
This item is not returnable.
View return policy
Description
FCHSD1 Polyclonal antibody specifically detects FCHSD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| FCHSD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| FCH and double SH3 domains 1, FCH and double SH3 domains protein 1, FLJ00007, nervous wreck homolog 2, NWK2 | |
| Produced in rabbits immunized with E. coli-derived Human FCHSD1 fragment | |
| 50 μg | |
| Neuroscience | |
| 89848 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction