missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCHSD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35232-20ul
This item is not returnable.
View return policy
Description
FCHSD1 Polyclonal antibody specifically detects FCHSD1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| FCHSD1 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FCH and double SH3 domains 1, FCH and double SH3 domains protein 1, FLJ00007, nervous wreck homolog 2, NWK2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human FCHSD1 (NP_258260.1).,, Sequence:, RLQASDRYRDLAGGTGRSAKEQVLRKGTENLQRAQAEVLQSVRELSRSRKLYGQRERVWALAQEKAADVQARLNRSDHGIFHSRTSLQKLSTKLSAQSAQYSQQLQAARNEYLLNLVATNAHLDHYYQEELPALLKALVSELSEHLRDPLTSLSHTELEAAEVILEHAHRGEQTTSQVSWEQDLKLFLQEPGVFSPTPPQQFQPAGTDQVCVLEWGAEGVAGKSGLEKEVQRLTSRAARDYKIQNHGHRVLQRLEQRRQQASEREAPSIEQRLQEVRESIRRAQVSQVKGAARLALLQGAG | |
| 20 μL | |
| Neuroscience | |
| 89848 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction