missing translation for 'onlineSavingsMsg'
Learn More

FCHSD1 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18400928 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400928 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400928 Supplier Novus Biologicals Supplier No. NBP222338R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FCHSD1 Polyclonal antibody specifically detects FCHSD1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCHSD1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FCH and double SH3 domains 1, FCH and double SH3 domains protein 1, FLJ00007, nervous wreck homolog 2, NWK2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 640 to 690 of human FCH and Double SH3 Domains 1 using the numbering given in entry NP_258260.1 (GeneID 89848).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 89848
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.