missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCGRT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552913
This item is not returnable.
View return policy
Description
Immunogen sequence: LTFLLFSCPH RLREHLERGR GNLEWKEPPS MRLKARPSSP GFSVLTCSAF SFYPPELQLR FLRNGLAAGT GQGDFGPNSD GSFHASSSLT VKSGDEHHYC CIVQHAGLAQ PLRVELESPA KS Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
FCGRT binds to the Fc region of monomeric immunoglobulins gamma. It mediates the uptake of IgG from milk. It plays a possible role in transfer of immunoglobulin G from mother to fetus.
Specifications
| FCGRT | |
| Polyclonal | |
| Unconjugated | |
| FCGRT | |
| alpha-chain; Fc fragment of IgG receptor and transporter; Fc fragment of IgG, receptor, transporter, alpha; Fc receptor, IgG, alpha chain transporter; FCGRT; FCRN; FcRn alpha chain; heavy chain of the major histocompatibility complex class I-like Fc receptor; IgG Fc fragment receptor transporter alpha chain; IgG receptor FcRn large subunit p51; immunoglobulin receptor, intestinal, heavy chain; major histocompatibility complex class I-like Fc receptor; neonatal Fc receptor; neonatal Fc-receptor for Ig; transmembrane alpha chain of the neonatal receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2217 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55899 | |
| FCGRT | |
| Recombinant protein corresponding to Human FCGRT. Recombinant protein control fragment (Product #RP-88667). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction