missing translation for 'onlineSavingsMsg'
Learn More

FcgR4/CD16-2 Antibody, FITC, Novus Biologicals™

Product Code. 18338256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338256 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338256 Supplier Novus Biologicals Supplier No. NBP305900F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FcgR4/CD16-2 Polyclonal specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcgR4/CD16-2
Applications ELISA
Classification Polyclonal
Conjugate FITC
Dilution ELISA
Formulation PBS
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse FcgR4/CD16-2 (NP_653142.2; Met 1-Gln 203;).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246256
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.