missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR3B/CD16b Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30201-1-AP-20UL
This item is not returnable.
View return policy
Description
The FCGR3B/CD16b antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FCGR3B/CD16b. This antibody recognizes human, rat antigen. The FCGR3B/CD16b antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| FCGR3B/CD16b | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2215 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 900 μg/mL | |
| CD16; CD16b; FCG3; FCGR3 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| 55 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction