Learn More
FCGR3A Mouse anti-Human, PE, Clone: COC16, Abnova™
Mouse monoclonal antibody raised against human FCGR3A.
Brand: Abnova MAB12395.50Reactions
Description
This gene encodes a receptor for the Fc portion of immunoglobulin G, and it is involved in the removal of antigen-antibody complexes from the circulation, as well as other other antibody-dependent responses. This gene (FCGR3A) is highly similar to another nearby gene (FCGR3B) located on chromosome 1. The receptor encoded by this gene is expressed on natural killer (NK) cells as an integral membrane glycoprotein anchored through a transmembrane peptide, whereas FCGR3B is expressed on polymorphonuclear neutrophils (PMN) where the receptor is anchored through a phosphatidylinositol (PI) linkage. Mutations in this gene have been linked to susceptibility to recurrent viral infections, susceptibility to systemic lupus erythematosus, and alloimmune neonatal neutropenia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| FCGR3A | |
| Monoclonal | |
| PE | |
| Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user. | |
| CD16/CD16A/FCG3/FCGR3/FCGRIII/FCR-10/FCRIII/FCRIIIA/IGFR3 | |
| Mouse | |
| Protein A/G affinity chromatography | |
| RUO | |
| 2214 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence | |
| COC16 | |
| Mouse monoclonal antibody raised against human FCGR3A. | |
| In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA) | |
| FCGR3A | |
| FCGR3A | |
| 50 Reactions | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.