missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR3A Mouse anti-Human, Clone: 3G8 (Biotin), Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13820.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human FCGR3A.
- High specificity
- High reactivity
Specifications
| FCGR3A | |
| Monoclonal | |
| Biotin | |
| Flow Cytometry (1 ug/106 cells) Functional Study Immunohistochemistry (Frozen sections) Immunoprecipitation The optimal working dilution should be determined by the end user. | |
| P08637 | |
| FCGR3A | |
| Human polymorphonuclear leukocytes. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation | |
| 3G8 | |
| Mouse monoclonal antibody raised against human FCGR3A. | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD16/CD16A/FCG3/FCGR3/FCGRIII/FCR-10/FCRIII/FCRIIIA/IGFR3 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 2214 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction