missing translation for 'onlineSavingsMsg'
Learn More

FCGR3A, 176 Phe/Val Recombinant Human Protein, His Tag

Product Code. 13432999
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13432999 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13432999 Supplier Invitrogen Supplier No. 10389H08C150

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

FCGR3A, 176 Phe/Val Recombinant Human Protein, His Tag

Fc fragment of IgG, low affinity IIIa, receptor (FCGR3A/CD16a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1- Gln 208) of human FCGR3A (P08637-1), (176 Phe/Val), to fused to a polyhistidine tag at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Assays and Analysis
Formulation Lyophilized
Molecular Weight (g/mol) 23.3kDa
Name FCGR3A 176 Phe/Val Recombinant Human Protein
Quantity 50 μg
Storage Requirements –20°C
Regulatory Status For research use only. Not for use in diagnostic procedures
Packaging 1 Each
Gene Alias RP11-5K23.1, CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Common Name FCGR3A 176 Phe/Val Recombinant Human Protein
Includes 1 Vial of lyophilized protein
Gene Symbol FCGR3A
Product Type FCGR3A 176 Phe/Val Recombinant Human Protein
Species Human
Protein Tag His Tag
Expression System Mammalian
Protein Family CD proteins, growth factors and receptors
Form Lyophilized
Protein Subtype Other proteins
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.