missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FCGR2C Polyclonal specifically detects FCGR2C in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | FCGR2C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | low affinity immunoglobulin gamma Fc region receptor II-c, CD32, CD32C, CDW32, Fc fragment of IgG, low affinity IIc, receptor for (CD32) (gene/pseudogene), FCG2, FCRIIC, IGFR2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 43-223 of human FCGR2C (NP_963857.3). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?