missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR2C Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | FCGR2C |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
FCGR2C Polyclonal specifically detects FCGR2C in Human samples. It is validated for Western Blot.Specifications
| FCGR2C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| low affinity immunoglobulin gamma Fc region receptor II-c, CD32, CD32C, CDW32, Fc fragment of IgG, low affinity IIc, receptor for (CD32) (gene/pseudogene), FCG2, FCRIIC, IGFR2 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human FCGR2C (NP_963857). Peptide sequence GTHSPESDSIPWFHNGNLIPTHTQPSYRFKANNNDSGEYTCQTGQTSLSD | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 9103 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit