Learn More
FCGR2A / CD32a Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27138-1-AP-150UL
 View more versions of this product
Product Code. 16875415
  • £349.00 / 150µL
Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
FCGR2A / CD32a
Polyclonal
Unconjugated
FCGR2A
CD32, CD32A, CDw32, Fc gamma RII a, Fc gamma RIIa, FCG2, FcGR, FCGR2, FCGR2A, FCGR2A1, FcRII a, IGFR2, IgG Fc receptor II a
Rabbit
Antigen Affinity Chromatography
RUO
2212
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12318
FCGR2A
FCGR2A / CD32a Fusion Protein Ag25878
150 μL