missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR1A, Mouse anti-Human, APC, Clone: 10.1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15364.100Reactions
This item is not returnable.
View return policy
Specifications
| FCGR1A | |
| Monoclonal | |
| APC | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD64/CD64A/FCRI/FLJ18345/IGFR1 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 10.1 | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| Fc fragment of IgG, high affinity Ia, receptor (CD64) | |
| FCGR1A | |
| Human rheumatoid synovial fluid cells and fibronectin-purified monocytes. | |
| RUO | |
| 2209 | |
| Store in the dark at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction