missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCER1A Mouse anti-Human, Clone:1F2A9, Abnova™
Mouse monoclonal antibody raised against recombinant human FCER1A.
Brand: Abnova MAB17840.100ug
This item is not returnable.
View return policy
Description
The IgE receptor plays a central role in allergic disease, coupling allergen and mast cell to initiate the inflammatory and immediate hypersensitivity responses that are characteristic of disorders such as hay fever and asthma. The allergic response occurs when 2 or more high-affinity IgE receptors are crosslinked via IgE molecules that in turn are bound to an allergen (antigen) molecule. A perturbation occurs that brings about the release of histamine and proteases from the granules in the cytoplasm of the mast cell and leads to the synthesis of prostaglandins and leukotrienes--potent effectors of the hypersensitivity response. The IgE receptor consists of 3 subunits: alpha, beta (MIM 147138), and gamma (MIM 147139); only the alpha subunit is glycosylated.[supplied by OMIM]Specifications
| FCER1A | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| FCER1A | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 1F2A9 | |
| Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide | |
| In PBS (0.05% sodium azide) | |
| FCE1A, FcERI | |
| Recombinant protein corresponding to amino acids 42-103 of human FCER1A from E. coli. | |
| 2205 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction