missing translation for 'onlineSavingsMsg'
Learn More

FCAR/CD89 Antibody (013), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18428229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428229 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428229 Supplier Novus Biologicals Supplier No. NBP290841JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FCAR/CD89 Monoclonal antibody specifically detects FCAR/CD89 in Rat samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCAR/CD89
Applications ELISA
Classification Monoclonal
Clone 013
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat FCAR/CD89 (Accession#: NP_973721.1; Met1-Asn228).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2204
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.