missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FCAMR Antibody (5B1D12E1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27130-0.025ml

 View more versions of this product

Product Code. 30027054

  • £226.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FCAMR Monoclonal antibody specifically detects FCAMR in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FCAMR
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
CD351 antigen, Fc alpha/mu receptor, Fc receptor, IgA, IgM, high affinity, FCA/MR, FKSG87, high affinity immunoglobulin alpha and immunoglobulin mu Fc receptor, immunity related factor, receptor for Fc fragment of IgA and IgM
Purified recombinant fragment of human FCAMR (AA: extra 221-450) expressed in E. Coli.
0.025 mL
Primary
Human
Purified
Flow Cytometry, ELISA, Western Blot
5B1D12E1
Unconjugated
PBS
Mouse
Protein G purified
RUO
83953
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.