missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIIA/CD16a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£190.00 - £255.00
Specifications
| Antigen | Fc gamma RIIIA/CD16a |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 μg/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629711
|
Novus Biologicals
NBP2-98147-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633764
|
Novus Biologicals
NBP2-98147-50ul |
50 μg |
£190.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIIIA/CD16a Polyclonal antibody specifically detects Fc gamma RIIIA/CD16a in Rat samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| Fc gamma RIIIA/CD16a | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 0.2 um filtered solution in PBS | |
| 2214 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 μg/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
| Produced in rabbits immunized with purified, recombinant Rat Fc gamma RIIIA/CD16a (Met 1-Pro 200; Accession#: ABO70343.1). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title