missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIB/CD32b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | Fc gamma RIIB/CD32b |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615305
|
Novus Biologicals
NBP2-99136-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623466
|
Novus Biologicals
NBP2-99136-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIIB/CD32b Polyclonal antibody specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for ELISASpecifications
| Fc gamma RIIB/CD32b | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| 0.2 um filtered solution in PBS | |
| 2213 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Produced in rabbits immunized with purified, recombinant Human Fc gamma RIIB/CD32b (Accession#: Ala 46-Pro 217; NP_001002274.1) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title