missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RIIB/CD32b Antibody (190907) [DyLight 405], Novus Biologicals™

Product Code. 30730943 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30730943 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730943 Supplier Novus Biologicals Supplier No. FAB14601E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Fc gamma RIIB/CD32b Monoclonal antibody specifically detects Fc gamma RIIB/CD32b in Mouse samples. It is validated for Western Blot,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc gamma RIIB/CD32b
Applications Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 190907
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIIB, aa 40-217, Accession # P08101
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2213
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.